Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

510351

Sigma-Aldrich

Davy Reagent methyl

technical grade

Synonym(s):

2,4-Bis(methylthio)-1,3,2,4-dithiadiphosphetane-2,4-disulfide, DR-Me

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6P2S6
CAS Number:
Molecular Weight:
284.41
Beilstein/REAXYS Number:
4182442
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

impurities

<15% p-dichlorobenzene

SMILES string

CSP1(=S)SP(=S)(SC)S1

InChI

1S/C2H6P2S6/c1-7-3(5)9-4(6,8-2)10-3/h1-2H3

InChI key

OIEQWZXDRGOGHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

105.8 °F

flash_point_c

41 °C

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service