Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

510319

Sigma-Aldrich

Piperidine tetrathiotungstate

technical grade

Synonym(s):

Tetrathiotungstic acid, compound with piperidine (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H24N2S4W
CAS Number:
Molecular Weight:
484.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

SMILES string

S[W](S)(=S)=S.C1CCNCC1.C2CCNCC2

InChI

1S/2C5H11N.2H2S.2S.W/c2*1-2-4-6-5-3-1;;;;;/h2*6H,1-5H2;2*1H2;;;/q;;;;;;+2/p-2

InChI key

SAHMQCBVHQHDMU-UHFFFAOYSA-L

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service