Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

510076

Sigma-Aldrich

Raney®-Copper

W.R. Grace and Co. Raney® 2900, slurry, in H2O, active catalyst

Synonym(s):

Copper sponge

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

slurry

manufacturer/tradename

W.R. Grace and Co. Raney® 2900

concentration

in H2O

SMILES string

[Cu]

Looking for similar products? Visit Product Comparison Guide

Legal Information

Raney is a registered trademark of W.R. Grace & Co.-Conn.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Åsa Björkbacka et al.
Dalton transactions (Cambridge, England : 2003), 44(36), 16045-16051 (2015-08-20)
One of the main challenges for the nuclear power industry today is the disposal of spent nuclear fuel. One of the most developed methods for its long term storage is the Swedish KBS-3 concept where the spent fuel is sealed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service