Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50365

Sigma-Aldrich

1,2,3-Trimethylimidazolium methyl sulfate

95%

Synonym(s):

Basionics® ST 99

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O4S
CAS Number:
Molecular Weight:
222.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥94.5% (HPLC)
95%

form

solid

impurities

≤0.5% water

anion traces

chloride (Cl-): ≤50 mg/kg

SMILES string

COS([O-])(=O)=O.Cc1n(C)cc[n+]1C

InChI

1S/C6H11N2.CH4O4S/c1-6-7(2)4-5-8(6)3;1-5-6(2,3)4/h4-5H,1-3H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

OUAUEIYYLHUEPK-UHFFFAOYSA-M

Legal Information

Basionics is a registered trademark of BASF SE

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

385.7 °F - closed cup

flash_point_c

196.50 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service