Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

498424

Sigma-Aldrich

3,5-Dimethylphenylzinc iodide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3ZnI
CAS Number:
Molecular Weight:
297.45
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Quality Level

concentration

0.5 M in THF

density

0.992 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Cc1cc(C)cc([Zn]I)c1

InChI

1S/C8H9.HI.Zn/c1-7-4-3-5-8(2)6-7;;/h4-6H,1-2H3;1H;/q;;+1/p-1

InChI key

KRLSFTGFMRQRTG-UHFFFAOYSA-M

Legal Information

Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

1.0 °F - closed cup

flash_point_c

-17.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service