498394
2,5-Dimethylphenylzinc iodide solution
0.5 M in THF
About This Item
Recommended Products
concentration
0.5 M in THF
bp
65 °C
density
1.004 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
Cc1ccc(C)c([Zn]I)c1
InChI
1S/C8H9.HI.Zn/c1-7-3-5-8(2)6-4-7;;/h3-5H,1-2H3;1H;/q;;+1/p-1
InChI key
KLJBMAZLRRWCKO-UHFFFAOYSA-M
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3 - Water-react 2
target_organs
Central nervous system, Respiratory system
supp_hazards
Storage Class
4.3 - Hazardous materials which set free flammable gases upon contact with water
wgk_germany
WGK 3
flash_point_f
1.4 °F - closed cup
flash_point_c
-17 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service