Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49822

Sigma-Aldrich

Tris[3-(heptadecafluorooctyl)phenyl]phosphine

≥95.0% (sum of phosphine, phosphine oxide, NMR)

Synonym(s):

Tris[3-(perfluorooctyl)phenyl]phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H12F51P
CAS Number:
Molecular Weight:
1516.44
Beilstein/REAXYS Number:
8824390
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥95.0% (sum of phosphine, phosphine oxide, NMR)

impurities

<10% phosphine oxide (NMR)

mp

88-93 °C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)c1cccc(c1)P(c2cccc(c2)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)c3cccc(c3)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Other Notes

Fluorinated phosphine used for synthesizing the Bannwarth catalyst for fluorous biphase palladium-catalyzed Suzuki couplings[1]; Stille coupling[2]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Schneider et al.
Helvetica Chimica Acta, 84, 735-735 (2001)
Repetitive Application of Perfluoro-Tagged Pd Complexes for Stille Couplings in a Fluorous Biphasic System This work was supported by Fluka (Buchs) and Byk Gulden Pharmaceuticals (Konstanz). S.S. thanks Fluka for financial support. We would also thank Priv.-Doz. Dr. C. Rücker for help in the preparation of the manuscript.
Schneider et al.
Angewandte Chemie (International ed. in English), 39(22), 4142-4145 (2000-11-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service