497258
Potassium thiocyanate
ReagentPlus®, ≥99.995%
Synonym(s):
Potassium rhodanide
About This Item
product line
ReagentPlus®
assay
≥99.995%
impurities
≤0.0005% insoluble matter
≤0.01% insol. Alcohol
<60 ppm total metallic impurities
pH
5.3-8.7
mp
173 °C (lit.)
anion traces
chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.005%
cation traces
Fe: ≤2 ppm
NH4+: ≤0.003%
Na: ≤0.005%
heavy metals: ≤5 ppm
SMILES string
[K]SC#N
InChI
1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1
InChI key
ZNNZYHKDIALBAK-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Other Notes
Legal Information
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service