Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

497053

Sigma-Aldrich

Merrifield′s peptide resin

100-200 mesh, extent of labeling: 2.0-3.0 mmol/g Cl- loading, 1 % cross-linked

Synonym(s):

Chloromethylated polystyrene, Chloromethylated stryene/divinylbenzene copolymer, Chloromethylated styrene-divinylbenzene resin, Copolymer of styrene and divinylbenzene, chloromethylated, Poly(styrene-co-vinylbenzyl chloride-co-divinylbenzene), Polystyrene crosslinked with divinylbenzene, chloromethylated

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

crosslinking

1 % cross-linked

extent of labeling

2.0-3.0 mmol/g Cl- loading

particle size

100-200 mesh

SMILES string

ClCc1ccccc1.C=Cc2ccccc2.C=Cc3ccc(C=C)cc3

InChI

1S/CH3Cl/c1-2/h1H3

InChI key

NEHMKBQYUWJMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service