Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

496782

Sigma-Aldrich

Hydroxypivalyl hydroxypivalate bis[6-(acryloyloxy)hexanoate]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2(CH2)5CO2CH2C(CH3)2CH2O2CC(CH3)2CH2O2C(CH2)5O2
CAS Number:
Molecular Weight:
540.64
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

contains

100 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.463 (lit.)

density

1.106 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(COC(=O)CCCCCOC(=O)C=C)COC(=O)C(C)(C)COC(=O)CCCCCOC(=O)C=C

InChI

1S/C28H44O10/c1-7-22(29)34-17-13-9-11-15-24(31)36-19-27(3,4)20-38-26(33)28(5,6)21-37-25(32)16-12-10-14-18-35-23(30)8-2/h7-8H,1-2,9-21H2,3-6H3

InChI key

HTMMMSIQFWMMIJ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service