490326
(1R,2S)-cis-1,2-Dihydro-1,2-naphthalenediol
0.067 g/mL in 0.1 M phosphate buffer, 98%
Synonym(s):
(1R-cis)-1,2-Dihydro-1,2-naphthalenediol, (1R,2S)-cis-1,2-Dihydro-1,2-dihydroxynaphthalene
About This Item
assay
98%
optical activity
[α]22/D +229°, c = 1 in methanol
concentration
0.067 g/mL in 0.1 M phosphate buffer
pH
8.5(adjusted to)
mp
121-127 °C (lit.)
SMILES string
O[C@H]1C=Cc2ccccc2[C@H]1O
InChI
1S/C10H10O2/c11-9-6-5-7-3-1-2-4-8(7)10(9)12/h1-6,9-12H/t9-,10+/m0/s1
InChI key
QPUHWUSUBHNZCG-VHSXEESVSA-N
Looking for similar products? Visit Product Comparison Guide
Physical form
Reconstitution
(1)Thaw the frozen suspension and filter the solid. (2) Rinse bottle with a few milliliters of base-washed (aqueous Na2CO3) ethyl acetate. (3) Use the rinsing to wash the solid. (4) Collect solid. (5) To further collect more product from the filtrate: extract filtrate with equal volumes of base-washed (aqueous Na2CO3) ethyl acetate (repeat three times). (6) Dry the ethyl acetate extract over MgSO4. (7) Evaporate the solvent - DO NOT HEAT! (8) Combine solids collected from the suspension and the filtrate. Pure crystals should be stored at -78° C. A suspension of the product in phosphate buffer is stable at 0° C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service