48921
3-(5-Carboxy-2H-benzotriazol-2-yl)-L-alanine
produced by Wacker Chemie AG, Burghausen, Germany, Wacker Chemie AG, ≥95.0% (HPLC)
Synonym(s):
(S)-2-Amino-3-(5-carboxy-2H-benzotriazol-2-yl)propionic acid
About This Item
grade
produced by Wacker Chemie AG, Burghausen, Germany
assay
≥95.0% (HPLC)
optical purity
ee: >99% (due to stereospecific enzymatic synthesis)
manufacturer/tradename
Wacker Chemie AG
storage temp.
2-8°C
SMILES string
N[C@@H](Cn1nc2ccc(cc2n1)C(O)=O)C(O)=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service