Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

485012

Sigma-Aldrich

Methacryloyl chloride

technical grade, 80%

Synonym(s):

α-Methylacryloyl chloride, 2-Methyl-2-propenoyl chloride, 2-Methylacryloyl chloride, 2-Methylpropenoic acid chloride, Methacrylic acid chloride, Methacrylic chloride, Methacrylyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COCl
CAS Number:
Molecular Weight:
104.53
Beilstein/REAXYS Number:
878175
EC Number:
MDL number:
UNSPSC Code:
12162002

grade

technical grade

assay

80%

contains

200 ppm phenothiazine as stabilizer

refractive index

n20/D 1.442 (lit.)

bp

95-96 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(Cl)=O

InChI

1S/C4H5ClO/c1-3(2)4(5)6/h1H2,2H3

InChI key

VHRYZQNGTZXDNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

55.0 °F - closed cup

flash_point_c

12.8 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service