Key Documents
482439
Sigma-Aldrich
ArgoGel-NH2
120 μm particle size
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352200
crosslinking
1 % cross-linked
extent of labeling
0.4-0.5 mmol N per g
particle size
120 μm
Legal Information
ArgoGel is a trademark of Argonaut Technologies, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)