Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

482366

Sigma-Aldrich

ArgoGel-OH

120 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

crosslinking

1 % cross-linked

extent of labeling

0.4-0.5 mmol OH per g

particle size

120 μm

SMILES string

OCCO.CC(COCCO)(COCCO)Cc1ccc(C=C)cc1

Legal Information

ArgoGel is a trademark of Argonaut Technologies, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service