About This Item
Recommended Products
assay
98%
mp
123-127 °C (lit.)
functional group
hydroxyl
nitro
SMILES string
OCc1cc(ccc1O)[N+]([O-])=O
InChI
1S/C7H7NO4/c9-4-5-3-6(8(11)12)1-2-7(5)10/h1-3,9-10H,4H2
Inchi Key
JEIYIXDNZKATLC-UHFFFAOYSA-N
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service