About This Item
assay
95%
refractive index
n20/D 1.523 (lit.)
bp
241 °C (lit.)
density
1.011 g/mL at 25 °C (lit.)
SMILES string
CCc1cccc(C(C)C)c1N=C=O
InChI
1S/C12H15NO/c1-4-10-6-5-7-11(9(2)3)12(10)13-8-14/h5-7,9H,4H2,1-3H3
InChI key
TVTUCESWFCLZQK-UHFFFAOYSA-N
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service