Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

477974

Sigma-Aldrich

N,N-Bis(2-acetoxyethyl)aniline

97%

Synonym(s):

N-Phenyldiethanolamine diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2CH2CH2)2NC6H5
CAS Number:
Molecular Weight:
265.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.523 (lit.)

bp

182 °C/1.2 mmHg (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OCCN(CCOC(C)=O)c1ccccc1

InChI

1S/C14H19NO4/c1-12(16)18-10-8-15(9-11-19-13(2)17)14-6-4-3-5-7-14/h3-7H,8-11H2,1-2H3

InChI key

XQGHEXBVXWBMGC-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service