Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

477109

Sigma-Aldrich

(R)-(+)-1,2-Epoxyoctane

97%

Synonym(s):

(R)-(+)-1-Octene oxide, (R)-(+)-2-Hexyloxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O
CAS Number:
Molecular Weight:
128.21
Beilstein/REAXYS Number:
79913
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

97%

optical activity

[α]25/D +14°, neat

refractive index

n20/D 1.42 (lit.)

bp

60-62 °C/15 mmHg (lit.)

density

0.839 g/mL at 25 °C (lit.)

SMILES string

CCCCCC[C@@H]1CO1

InChI

1S/C8H16O/c1-2-3-4-5-6-8-7-9-8/h8H,2-7H2,1H3/t8-/m1/s1

InChI key

NJWSNNWLBMSXQR-MRVPVSSYSA-N

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

98.6 °F - closed cup

flash_point_c

37 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service