About This Item
Recommended Products
assay
97%
mp
45-48 °C (lit.)
SMILES string
Ic1cccc(c1)N=C=S
InChI
1S/C7H4INS/c8-6-2-1-3-7(4-6)9-5-10/h1-4H
InChI key
LKCSXVAACHUTGC-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Sens. 1
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service