Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47571

Sigma-Aldrich

Fmoc-Glu(OBzl)-OH

≥98.0% (HPLC)

Synonym(s):

Fmoc-L-glutamic acid 5-benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H25NO6
CAS Number:
Molecular Weight:
459.49
Beilstein/REAXYS Number:
3576722
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

assay

≥98.0% (HPLC)

optical activity

[α]20/D +12.0±2.5°, c = 1% in chloroform

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](CCC(=O)OCc1ccccc1)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C27H25NO6/c29-25(33-16-18-8-2-1-3-9-18)15-14-24(26(30)31)28-27(32)34-17-23-21-12-6-4-10-19(21)20-11-5-7-13-22(20)23/h1-13,23-24H,14-17H2,(H,28,32)(H,30,31)/t24-/m0/s1

InChI key

HJJURMMMGPQIQP-DEOSSOPVSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Bodkowski et al.
Animals : an open access journal from MDPI, 10(10) (2020-10-15)
The study aimed to investigate the effect of supplementation of fish oil after the process of low-temperature crystallization (LTC-FO) enriched with long-chain polyunsaturated fatty acids (LC-PUFAs) on cow milk parameters. The experiment was carried out on 24 Polish Holstein Friesian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service