Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47418

Sigma-Aldrich

Fmoc-β-(3-benzothienyl)-Ala-OH

≥98.0% (HPLC)

Synonym(s):

Fmoc-3-(3-benzothienyl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H21NO4S
CAS Number:
Molecular Weight:
443.51
Beilstein/REAXYS Number:
9296209
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98.0% (HPLC)

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1csc2ccccc12)NC(=O)OCC3c4ccccc4-c5ccccc35

Looking for similar products? Visit Product Comparison Guide

Linkage

Thia analog of FMOC-L-Tryptophan

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service