Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

473243

Sigma-Aldrich

8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt

Dye content 75 %

Synonym(s):

HPTS, Pyranine, Solvent Green 7, Trisodium 8-hydroxypyrene-1,3,6-trisulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H7Na3O10S3
CAS Number:
Molecular Weight:
524.39
Colour Index Number:
59040
Beilstein/REAXYS Number:
4107272
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

composition

Dye content, 75%

mp

>300 °C (lit.)

λmax

403 nm

SMILES string

[Na+].[Na+].[Na+].Oc1cc(c2ccc3c(cc(c4ccc1c2c34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H10O10S3.3Na/c17-11-5-12(27(18,19)20)8-3-4-10-14(29(24,25)26)6-13(28(21,22)23)9-2-1-7(11)15(8)16(9)10;;;/h1-6,17H,(H,18,19,20)(H,21,22,23)(H,24,25,26);;;/q;3*+1/p-3

InChI key

KXXXUIKPSVVSAW-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

481.3 °F

flash_point_c

249.6 °C

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felix H C Wong et al.
Journal of fluorescence, 21(1), 299-312 (2010-10-06)
Steep variations in concentration and temperature frequently occur in small fluid compartments such as those found in cells or microfluidic devices. A quantitative characterization of concentration and temperature gradients is therefore required before these systems can be fully understood. Although
Takashi Kikukawa et al.
Biochemistry, 50(41), 8888-8898 (2011-09-13)
Acetabularia rhodopsins are the first microbial rhodopsins discovered in a marine plant organism, Acetabularia acetabulum. Previously, we expressed Acetabularia rhodopsin II (ARII) by a cell-free system from one of two opsin genes in A. acetabulum cDNA and showed that ARII
Tridib Mondal et al.
The journal of physical chemistry. B, 114(41), 13136-13142 (2010-09-25)
Excited state proton transfer (ESPT) of pyranine (8-hydroxypyranine-1,3,6-trisulfonate, HPTS) in room temperature ionic liquid (RTIL) mixed micelles is studied by femtosecond up-conversion. The mixed micelle consists of a triblock copolymer, (PEO)(20)-(PPO)(70)-(PEO)(20) (Pluronic P123), and one of the two RTILs, 1-pentyl-3-methyl-imidazolium
Boiko Cohen et al.
The journal of physical chemistry. B, 115(23), 7637-7647 (2011-04-12)
We report on femto- to nanosecond studies of the excited state intermolecular proton transfer (ESPT) reaction of trisodium 8-hydroxypyrene-1,3,6-trisulfonate (pyranine, HPTS) with the human serum albumin (HSA) protein. The formed robust 1:1 complexes (K(eq) = (2.6 ± 0.1) × 10(6)
Patrick Maurer et al.
The Journal of chemical physics, 133(4), 044108-044108 (2010-08-07)
Ultrafast, time-resolved investigations of acid-base neutralization reactions have recently been performed using systems containing the photoacid 8-hydroxypyrene-1,3,6-trisulfonic acid trisodium salt (HPTS) and various Bronsted bases. Two conflicting neutralization mechanisms have been formulated by Mohammed et al. [Science 310, 83 (2005)]

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service