Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

472948

Sigma-Aldrich

Ytterbium(III) chloride hexahydrate

99.99%

Synonym(s):

Ytterbium trichloride hydrate, Ytterbium(III) chloride·6hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YbCl3 · 6H2O
CAS Number:
Molecular Weight:
387.49
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

assay

99.99%

mp

180 °C (dec.) (lit.)

density

2.58 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Yb](Cl)Cl

InChI

1S/3ClH.6H2O.Yb/h3*1H;6*1H2;/q;;;;;;;;;+3/p-3

InChI key

LEYFXTUKPKKWMP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uliana Kostiv et al.
Beilstein journal of nanotechnology, 6, 2290-2299 (2016-01-07)
NaYF4:Yb(3+)/Er(3+) nanoparticles were synthesized by thermal decomposition of lanthanide trifluoroacetates using oleylamine (OM) as both solvent and surface binding ligand. The effect of reaction temperature and time on the properties of the particles was investigated. The nanoparticles were characterized by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service