Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

471534

Sigma-Aldrich

Methanesulfonamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2NH2
CAS Number:
Molecular Weight:
95.12
Beilstein/REAXYS Number:
1740835
EC Number:
MDL number:
UNSPSC Code:
12352100

Quality Level

assay

97%

mp

85-89 °C (lit.)

SMILES string

CS(N)(=O)=O

InChI

1S/CH5NO2S/c1-5(2,3)4/h1H3,(H2,2,3,4)

InChI key

HNQIVZYLYMDVSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Arkema Inc.

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service