470848
Lithium diisopropylamide
30 wt. % suspension in mineral oil
Synonym(s):
LDA
About This Item
vapor density
>1 (vs air)
concentration
30 wt. % suspension in mineral oil
density
0.864 g/mL at 25 °C (lit.)
SMILES string
[Li]N(C(C)C)C(C)C
InChI
1S/C6H14N.Li/c1-5(2)7-6(3)4;/h5-6H,1-4H3;/q-1;+1
Inchi Key
ZCSHNCUQKCANBX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
91.4 °F - closed cup
flash_point_c
33 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service