Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

470732

Sigma-Aldrich

Antimony(III) isopropoxide

99.99% trace metals basis

Synonym(s):

Antimony triisopropoxide, Antimony triisopropylate, Antimony(III) isopropoxide, Triisopropoxyantimony

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sb(OCH(CH3)2)3
CAS Number:
Molecular Weight:
299.02
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

99.99% trace metals basis

form

liquid

reaction suitability

core: antimony
reagent type: catalyst

bp

53-54 °C/0.25 mmHg (lit.)

density

1.307 g/mL at 25 °C (lit.)

SMILES string

CC(C)O[Sb](OC(C)C)OC(C)C

InChI

1S/3C3H7O.Sb/c3*1-3(2)4;/h3*3H,1-2H3;/q3*-1;+3

Inchi Key

HYPTXUAFIRUIRD-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

170.6 °F - closed cup

flash_point_c

77 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service