Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

469637

Sigma-Aldrich

N-Benzylidene-tert-butylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=NC(CH3)3
CAS Number:
Molecular Weight:
161.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.52 (lit.)

bp

209 °C (lit.)

density

0.906 g/mL at 25 °C (lit.)

functional group

amine
imine
phenyl

SMILES string

CC(C)(C)\N=C\c1ccccc1

InChI

1S/C11H15N/c1-11(2,3)12-9-10-7-5-4-6-8-10/h4-9H,1-3H3/b12-9+

InChI key

KFLSWDVYGSSZRX-FMIVXFBMSA-N

General description

In vitro metabolism of N-benzyl-tert-butylamine in male hamster hepatic microsomal preparations has been studied.[1] Kinetic studies of frustrated Lewis pair (FLP)-catalyzed hydrogenation and deuteration of N-benzylidene-tert-butylamine suggests that the reaction proceeds via autoinductive catalysis.[2]

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

172.4 °F - closed cup

flash_point_c

78 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sebastian Tussing et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(22), 8056-8059 (2015-04-17)
The frustrated Lewis pair (FLP)-catalyzed hydrogenation and deuteration of N-benzylidene-tert-butylamine (2) was kinetically investigated by using the three boranes B(C6F5)3 (1), B(2,4,6-F3-C6H2)3 (4), and B(2,6-F2-C6H3)3 (5) and the free activation energies for the H2 activation by FLP were determined. Reactions
M Ulgen et al.
Drug metabolism and drug interactions, 12(2), 131-143 (1995-01-01)
The metabolism of N-benzyl-tert-butylamine was studied in vitro using male hamster hepatic microsomal preparations. This substrate produced the corresponding nitrone, benzaldehyde and an uncharacterised metabolite. No metabolites were detected which corresponded to either authentic amide or oxaziridine. The results indicate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service