Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

467774

Sigma-Aldrich

Sodium nitrate

≥99.99%, ReagentPlus®

Synonym(s):

Chile salpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaNO3
CAS Number:
Molecular Weight:
84.99
EC Number:
MDL number:
UNSPSC Code:
12352302

product line

ReagentPlus®

assay

≥99.99%

impurities

≤0.005% insolubles
<100 ppm total metallic impurities

pH

5.5-8.3

mp

306 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.001%
nitrite (NO2-): ≤0.001%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.003%

cation traces

Ca, Mg and R2O3 precipitate: ≤0.005%
Fe: ≤3 ppm
heavy metals: ≤5 ppm

SMILES string

[Na+].[O-][N+]([O-])=O

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

pictograms

Flame over circleExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suna Choi et al.
Journal of controlled release : official journal of the Controlled Release Society, 131(1), 70-76 (2008-08-12)
One of the most important requirements for non-viral gene delivery systems is the ability to mediate high levels of gene expression with low toxicity. After the DNA/vector complexes are taken up by cells through endocytosis, DNA is typically contained within
Filip J Larsen et al.
The American journal of clinical nutrition, 99(4), 843-850 (2014-02-07)
Nitrate, which is an inorganic anion abundant in vegetables, increases the efficiency of isolated human mitochondria. Such an effect might be reflected in changes in the resting metabolic rate (RMR) and formation of reactive oxygen species. The bioactivation of nitrate
Daryoush Afzali et al.
Journal of chromatography. A, 1247, 35-41 (2012-06-08)
In the present study, a new approach which uses immunoaffinity column clean-up combined with dispersive liquid-liquid microextraction (DLLME) is proposed for the preconcentration of ultra trace amounts of aflatoxins (B₁, B₂, G₁ and G₂). The aflatoxins are then determined using
Raúl Bescós et al.
Medicine and science in sports and exercise, 44(12), 2400-2409 (2012-07-20)
Supplementation with inorganic nitrate has been suggested to be an ergogenic aid for athletes as nitric oxide donor. The purpose of this study was to determine whether ingestion of inorganic sodium nitrate benefits well-trained athletes performing a 40-min exercise test
Ibrahim Yazji et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(23), 9451-9456 (2013-05-08)
Necrotizing enterocolitis (NEC) is a devastating disease of premature infants characterized by severe intestinal necrosis and for which breast milk represents the most effective protective strategy. Previous studies have revealed a critical role for the lipopolysaccharide receptor toll-like receptor 4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service