467499
[1-(4-Morpholinyl)ethyl]benzotriazole, mixture of Bt1 and Bt2 isomers
About This Item
reaction suitability
reaction type: C-C Bond Formation
impurities
0.5% ethyl alcohol
10% benzotriazole
SMILES string
C1CN(CCO1)CCn2nnc3ccccc23
InChI
1S/C12H16N4O/c1-2-4-12-11(3-1)13-14-16(12)6-5-15-7-9-17-10-8-15/h1-4H,5-10H2
InChI key
ZWPLZUXFENZGRI-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
4.1B - Flammable solid hazardous materials
wgk_germany
WGK 3
flash_point_f
134.6 °F - closed cup
flash_point_c
57 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service