Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

467197

Sigma-Aldrich

4-Methoxybenzyl phenylacetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2CH2C6H4OCH3
CAS Number:
Molecular Weight:
256.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.559 (lit.)

bp

370 °C (lit.)

solubility

organic solvents: soluble(lit.)

density

1.128 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(COC(=O)Cc2ccccc2)cc1

InChI

1S/C16H16O3/c1-18-15-9-7-14(8-10-15)12-19-16(17)11-13-5-3-2-4-6-13/h2-10H,11-12H2,1H3

InChI key

VCYWCSZLXMMLLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service