Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

463124

Sigma-Aldrich

Phosphine-d3

electronic grade, 98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PD3
CAS Number:
Molecular Weight:
37.02
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

electronic grade

isotopic purity

98 atom % D

form

gas

bp

−87.5 °C (lit.)

mass shift

M+3

SMILES string

[2H]P([2H])[2H]

Recommended products

Stainless steel regulators Z527416 or Z527424 are recommended.

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Liquefied gas - Self-heat. 1 - Skin Corr. 1B

Storage Class

2A - Gases

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service