460842
Sulfasalazine
97%
Synonym(s):
2-Hydroxy-5-{{4-[(2-pyridinylamino)sulfonyl]phenyl}azo}benzoic acid, 5-[4-(2-Pyridylsulfamoyl)phenylazo]salicylic acid, Salicylazosulfapyridine
About This Item
Recommended Products
assay
97%
mp
260-265 °C (dec.) (lit.)
SMILES string
OC(=O)c1cc(ccc1O)\N=N\c2ccc(cc2)S(=O)(=O)Nc3ccccn3
InChI
1S/C18H14N4O5S/c23-16-9-6-13(11-15(16)18(24)25)21-20-12-4-7-14(8-5-12)28(26,27)22-17-3-1-2-10-19-17/h1-11,23H,(H,19,22)(H,24,25)/b21-20+
InChI key
NCEXYHBECQHGNR-QZQOTICOSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service