Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

460184

Sigma-Aldrich

Tri(ethylene glycol) bis(2-ethylhexanoate)

average Mn ~374, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3CH(C2H5)CO2CH2CH2OCH2-]2
CAS Number:
Molecular Weight:
402.57
EC Number:
MDL number:
UNSPSC Code:
12162002

grade

technical grade

mol wt

average Mn ~374

impurities

10% polyalkyl substitution groups

refractive index

n20/D 1.445

bp

344 °C (lit.)

mp

−50 °C (lit.)

solubility

H2O: insoluble
hexane, toluene, ethanol and acetone: soluble

density

0.97 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)C(=O)OCCOCCOCCOC(=O)C(CC)CCCC

InChI

1S/C22H42O6/c1-5-9-11-19(7-3)21(23)27-17-15-25-13-14-26-16-18-28-22(24)20(8-4)12-10-6-2/h19-20H,5-18H2,1-4H3

InChI key

FRQDZJMEHSJOPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer

Packaging

Packaged in glass bottles

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service