Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

459887

Sigma-Aldrich

L-Aspartic acid dibenzyl ester p-toluenesulfonate salt

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCOCH2CH(NH2)COOCH2C6H5 · C7H8O3S
CAS Number:
Molecular Weight:
485.55
Beilstein/REAXYS Number:
3647510
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

98%

optical activity

[α]24/D +9°, c = 1 in chloroform

mp

157-160 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cc1ccc(cc1)S(O)(=O)=O.N[C@@H](CC(=O)OCc2ccccc2)C(=O)OCc3ccccc3

InChI

1S/C18H19NO4.C7H8O3S/c19-16(18(21)23-13-15-9-5-2-6-10-15)11-17(20)22-12-14-7-3-1-4-8-14;1-6-2-4-7(5-3-6)11(8,9)10/h1-10,16H,11-13,19H2;2-5H,1H3,(H,8,9,10)/t16-;/m0./s1

InChI key

HLMUYZYLPUHSNV-NTISSMGPSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service