Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

453390

Sigma-Aldrich

(1R,2S)-(−)-trans-2-(1-Methyl-1-phenylethyl)cyclohexanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(CH3)2C6H10OH
CAS Number:
Molecular Weight:
218.33
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

assay

97%

optical activity

[α]24/D −29°, c = 1.7 in methanol

refractive index

n20/D 1.541 (lit.)

bp

300 °C (lit.)

density

1.026 g/mL at 25 °C (lit.)

SMILES string

CC(C)([C@@H]1CCCC[C@H]1O)c2ccccc2

InChI

1S/C15H22O/c1-15(2,12-8-4-3-5-9-12)13-10-6-7-11-14(13)16/h3-5,8-9,13-14,16H,6-7,10-11H2,1-2H3/t13-,14-/m1/s1

InChI key

CTYOSJAXQPHWCK-ZIAGYGMSSA-N

Application

Cyclohexyl-based chiral auxiliary that provides equal or greater levels of asymmetric induction when compared to menthol or 8-phenylmenthol in a variety of reactions.[1][2][3][4]

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comins, D.L. et al.
Journal of the American Chemical Society, 116, 4719-4719 (1994)
Comins, D.L. Guerra-Weltzien, L.
Tetrahedron Letters, 37, 3807-3807 (1996)
Comins, D.L. Radi Benjelloun, N.
Tetrahedron Letters, 35, 829-829 (1994)
Comins, D.L. et al.
Synlett, 972-972 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service