Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

453226

Sigma-Aldrich

(+)-Corey lactone, 4-phenylbenzoate alcohol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O5
CAS Number:
Molecular Weight:
352.38
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

assay

99%

optical activity

[α]20/D +87°, c = 1 in chloroform

mp

134-136 °C (lit.)

SMILES string

OC[C@@H]1[C@H](C[C@H]2OC(=O)C[C@@H]12)OC(=O)c3ccc(cc3)-c4ccccc4

Application

Key intermediate in the synthesis of prostaglandins.[1][2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 15, 65-65 (1982)
B Resul et al.
Journal of medicinal chemistry, 36(2), 243-248 (1993-01-22)
A series of phenyl-substituted analogues of prostaglandin F2 alpha (PGF2 alpha) were prepared and evaluated for ocular hypotensive effect and side effects in different animal models. In addition, the activity of the analogues on FP receptors was studied in vitro.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service