Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

451452

Sigma-Aldrich

N-Methyltrimethylacetamide

98%

Synonym(s):

N,2,2-Trimethylpropionamide, N-Methylpivalamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCONHCH3
CAS Number:
Molecular Weight:
115.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

mp

86-91 °C (lit.)

SMILES string

CNC(=O)C(C)(C)C

InChI

1S/C6H13NO/c1-6(2,3)5(8)7-4/h1-4H3,(H,7,8)

InChI key

QMKKJBRRKIKWFK-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation of nylon model systems.
Burnett GM and Riches KM.
Journal of the Chemical Society B: Physical Organic, 1229-1234 (1966)
Lipase-catalyzed transamidation of non-activated amides in organic solvent.
Sergeeva MV, et al.
Biotechnology Letters, 22(17), 1419-1422 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service