Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

451037

Sigma-Aldrich

Iodine

beads, anhydrous, −60 mesh, 99.999%

Synonym(s):

Diiodine, Iodine crystals, Molecular iodine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
Beilstein/REAXYS Number:
3587194
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

anhydrous

vapor density

9 (vs air)

vapor pressure

0.31 mmHg ( 25 °C)
1 mmHg ( 38.7 °C)

assay

99.999%

form

beads

resistivity

1.3E15 μΩ-cm

particle size

−60 mesh

bp

184 °C (lit.)

mp

113 °C (lit.)

SMILES string

II

InChI

1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service