Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

450766

Sigma-Aldrich

Cesium bromide

anhydrous, beads, −10 mesh, 99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsBr
CAS Number:
Molecular Weight:
212.81
EC Number:
MDL number:
UNSPSC Code:
12352302

grade

anhydrous

assay

99.9% trace metals basis

form

beads

particle size

−10 mesh

mp

636 °C (lit.)

density

4.44 g/mL at 25 °C (lit.)

SMILES string

[Br-].[Cs+]

InChI

1S/BrH.Cs/h1H;/q;+1/p-1

InChI key

LYQFWZFBNBDLEO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Chen Zheng et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 64(3), 628-630 (2006-01-03)
The complete high-order perturbation formula of g factor, including not only the widely used crystal-field (CF) mechanism, but also the neglected change-transfer (CT) mechanism in the CF theory, is established for a 3d8 ion in cubic octahedral site. From the
N Devaraj et al.
Analytical biochemistry, 206(1), 142-146 (1992-10-01)
A procedure for the rapid isolation of mucin glycoprotein by density gradient centrifugation in cesium trifluoroacetate (CsTFA) is described. The separation of mixtures of rat tracheobronchial mucin, DNA, hyaluronic acid, and bovine serum albumin in CsTFA gradients was superior to
Caesium-binding sites in the gramicidin pore.
D A Doyle et al.
Biochemical Society transactions, 22(4), 1043-1045 (1994-11-01)
Peng Pan et al.
Analytical chemistry, 75(20), 5468-5474 (2004-01-09)
Solutions consisting of protein and small molecule mixtures have been subjected to electrospray ionization to study the influence of small molecule/cation components at high concentrations on the electrospray responses of proteins. Emphasis was placed on solutions consisting of equal parts
N Kurata et al.
Radiation protection dosimetry, 119(1-4), 398-401 (2006-06-01)
CsBr phosphor ceramics doped with different luminescence centres such as In2O3, Eu2O3, EuCl3, SmCl3, TbCl3, GdCl3 or NdCl3 as the candidate for a new optically-stimulable phosphor for medical X-ray imaging sensor were prepared using a conventional ceramic fabrication process. It

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service