450472
4,4′-(1,4-Phenylenediisopropylidene)bisphenol
99%
Synonym(s):
Bisphenol P
About This Item
Recommended Products
assay
99%
mp
193-195 °C (lit.)
SMILES string
CC(C)(c1ccc(O)cc1)c2ccc(cc2)C(C)(C)c3ccc(O)cc3
InChI
1S/C24H26O2/c1-23(2,19-9-13-21(25)14-10-19)17-5-7-18(8-6-17)24(3,4)20-11-15-22(26)16-12-20/h5-16,25-26H,1-4H3
InChI key
GIXXQTYGFOHYPT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
pcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service