Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

449695

Sigma-Aldrich

Cadmium iodide

anhydrous, beads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Cadmium diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CdI2
CAS Number:
Molecular Weight:
366.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

assay

99.999% trace metals basis

form

beads

reaction suitability

reagent type: catalyst
core: cadmium

impurities

≤20.0 ppm Trace Metal Analysis

particle size

−10 mesh

bp

787 °C (lit.)

mp

388 °C (lit.)

density

5.67 g/mL at 25 °C (lit.)

SMILES string

I[Cd]I

InChI

1S/Cd.2HI/h;2*1H/q+2;;/p-2

InChI key

OKIIEJOIXGHUKX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S V Myl'nikov
TSitologiia i genetika, 25(4), 67-72 (1991-07-01)
In prolonged genetic experiment two contrast inbred strains, selected for the differences in embryonic mortality have been produced (HEM and LEM). In HEM strain the balanced lethal system in 55.0 region (near B1) of chromosome 2 has been formed. This
Haitao Guo et al.
Optics express, 17(17), 15350-15358 (2009-08-19)
Two serial Dy(3+)-doped and Dy(3+), Tm(3+)-codoped (100-x)(0.8GeS(2).0.2Ga(2)S(3)).xCdI(2) (0<or=x<or=20) chalcohalide glasses were prepared and characterized. By analyzing the absorption and emission measurements of the two serial chalcohalide glasses with the Judd-Ofelt analysis, we were able to calculate its Judd-Ofelt strength parameters
D V Zhuravleva et al.
Bioorganicheskaia khimiia, 22(10-11), 870-874 (1996-10-01)
Using cytochrome c from horse heart and the yeast candida valida as examples, it was shown that a complex anion, cadmium tetraiodide (CdI42-), precipitated proteins from aqueous solutions at the reagent concentrations below 50 mM. The composition and pH value
M Idrish Miah
The journal of physical chemistry. B, 113(6), 1652-1654 (2009-01-23)
Photoinduced second harmonic generation (PISHG) in undoped as well as in various Cu-doped (0.05-1.2% Cu) CdI2 nanocrystals was measured at liquid nitrogen temperature (LNT). It was found that the PISHG increases with increasing Cu doping up to approximately 0.6% and
Zheng-Bo Han et al.
Inorganic chemistry, 49(3), 769-771 (2010-01-06)
The anionic CdI(2)-type topological net, [Cu(2)(tci)(2)](2-), and the pentanuclear copper cluster cation [Cu(5)(tci)(2)(OH)(2)(H(2)O)(8)](2+) [tci = tris(2-carboxyethyl)isocyanurate] form a complementary 3D supramolecular framework. Interestingly, there exist centrosymmetric cyclic (H(2)O)(18) clusters in the cavities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service