Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

449687

Sigma-Aldrich

Cadmium chloride

anhydrous, beads, −10 mesh, ≥99.9%

Synonym(s):

Cadmium dichloride, Dichlorocadmium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CdCl2
Molecular Weight:
183.32
EC Number:
MDL number:
UNSPSC Code:
12352302

grade

anhydrous

vapor density

6.3 (vs air)

vapor pressure

10 mmHg ( 656 °C)

assay

≥99.9%

form

beads

particle size

−10 mesh

bp

964 °C/1 atm (lit.)

mp

568 °C (lit.)

solubility

acetone: slightly soluble(lit.)
diethyl ether: insoluble(lit.)
ethanol: slightly soluble(lit.)

SMILES string

Cl[Cd]Cl

InChI

1S/Cd.2ClH/h;2*1H/q+2;;/p-2

InChI key

YKYOUMDCQGMQQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa Coccini et al.
Histology and histopathology, 28(2), 195-209 (2013-01-01)
Potential risk associated with new nanomaterial exposure needs to be assessed. This in vivo study investigated pulmonary effects of engineered cadmium-containing silica nanoparticles Cd/SiNPs (1 mg/rat), silica SiNPs (600 μg/rat) and CdCl₂ (400 μg/rat) 1, 7 and 30 days after
Michael D Menz et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(10), 4550-4560 (2013-03-08)
Focused ultrasound is a promising noninvasive technology for neural stimulation. Here we use the isolated salamander retina to characterize the effect of ultrasound on an intact neural circuit and compared these effects with those of visual stimulation of the same
Veronique E Miron et al.
Nature neuroscience, 16(9), 1211-1218 (2013-07-23)
The lack of therapies for progressive multiple sclerosis highlights the need to understand the regenerative process of remyelination that can follow CNS demyelination. This involves an innate immune response consisting of microglia and macrophages, which can be polarized to distinct
M Auslander et al.
Marine biotechnology (New York, N.Y.), 10(3), 250-261 (2008-01-24)
Individuals of the fish Lithognathus mormyrus were exposed to a series of pollutants including: benzo[a]pyrene, pp-DDE, Aroclor 1254, perfluorooctanoic acid, tributyl-tin chloride, lindane, estradiol, 4-nonylphenol, methyl mercury chloride, and cadmium chloride. Five mixtures of the pollutants were injected. Each mixture
Grace Choong et al.
Toxicology and applied pharmacology, 272(2), 423-430 (2013-07-23)
Cadmium disrupts the actin cytoskeleton in rat mesangial cells, and we have previously shown that this involves a complex interplay involving activation of kinase signaling, protein translocation, and disruption of focal adhesions. Here we investigate the role that glutathionylation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service