Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

446661

Sigma-Aldrich

2,5-Dimethylbenzenethiol

90%, technical grade

Synonym(s):

2,5-Dimethylthiophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3SH
CAS Number:
Molecular Weight:
138.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical grade

assay

90%

refractive index

n20/D 1.5698 (lit.)

bp

126.3 °C/50 mmHg (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)c(S)c1

InChI

1S/C8H10S/c1-6-3-4-7(2)8(9)5-6/h3-5,9H,1-2H3

InChI key

NHAUBUMQRJWWAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service