Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

446599

Sigma-Aldrich

1,2-Dibromohexane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(Br)CH2Br
CAS Number:
Molecular Weight:
243.97
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.503 (lit.)

bp

89-90 °C/18 mmHg (lit.)

density

1.577 g/mL at 25 °C (lit.)

SMILES string

CCCCC(Br)CBr

InChI

1S/C6H12Br2/c1-2-3-4-6(8)5-7/h6H,2-5H2,1H3

InChI key

NUUUANNAZVEWHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New C~ 2-Fragment for Ruthenium-Catalyzed Synthesis of Indoles.
Cho CS, et al.
Bull. Korean Chem. Soc., 28(5), 832-832 (2007)
Synthesis of 2, 2-Dialkylcyclopropane Nitriles1.
Nelson ER, et al.
Journal of the American Chemical Society, 79(13), 3467-3469 (1957)
Cyclodextrins as chiral stationary phases in capillary gas chromatography. Part IV: Heptakis (2, 3, 6-tri-O-pentyl)-?-cyclodextrin.
Konig WA, et al.
Journal of High Resolution Chromatography, 12(1), 35-39 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service