Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

445665

Sigma-Aldrich

Tetrakis(dimethylamido)zirconium(IV)

crystalline

Synonym(s):

TDMAZ, Tetrakis(dimethylamino)zirconium(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]4Zr
CAS Number:
Molecular Weight:
267.53
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

crystalline
solid

reaction suitability

core: zirconium
reagent type: catalyst

mp

57-60 °C (lit.)

SMILES string

CN(C)[Zr](N(C)C)(N(C)C)N(C)C

InChI

1S/4C2H6N.Zr/c4*1-3-2;/h4*1-2H3;/q4*-1;+4

InChI key

DWCMDRNGBIZOQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

target_organs

Respiratory system

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service