Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

444855

Sigma-Aldrich

Acetone-d6

99.9 atom % D

Synonym(s):

Hexadeuteroacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3COCD3
CAS Number:
Molecular Weight:
64.12
Beilstein/REAXYS Number:
1702935
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

vapor density

2 (vs air)

Quality Level

vapor pressure

14.39 psi ( 55 °C)
184 mmHg ( 20 °C)
3.67 psi ( 20 °C)

isotopic purity

99.9 atom % D

expl. lim.

13.2 %

packaging

pk of 10 × 0.75 mL

refractive index

n20/D 1.355 (lit.)

bp

55.5 °C (lit.)

mp

−93.8 °C (lit.)

density

0.872 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])C(=O)C([2H])([2H])[2H]

InChI

1S/C3H6O/c1-3(2)4/h1-2H3/i1D3,2D3

InChI key

CSCPPACGZOOCGX-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-2.2 °F - closed cup

flash_point_c

-19 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Busseron et al.
ACS nano, 9(3), 2760-2772 (2015-03-04)
A family of triarylamine-fullerene conjugates has been synthesized and shown to self-assemble upon light stimulation in chlorinated solvents. This light-induced process primarily involves excitation of triarylamine derivatives, which then oxidize and stack with their neutral counterparts to form charge transfer
Ofentse Mazimba et al.
Bioorganic & medicinal chemistry, 22(23), 6564-6569 (2014-12-03)
A series of 3-nitrochalcones have been synthesized enroute towards fused ring pyrazolones and isoxazolones. Base catalyzed condensation of the chalcones with ethylacetoacetate yielded cyclohexenones in good yields (74-76%). The treatment of cyclohexenones with hydrazine hydrate or hydroxylamine chloride in the
Anna K F Mårtensson et al.
Dalton transactions (Cambridge, England : 2003), 44(8), 3604-3613 (2014-11-20)
Linear and circular dichroism (LD and CD) spectroscopy as well as isothermal titration calorimetry (ITC) have been used to investigate the interaction of Ru(tpy)(py)dppz(2+) (tpy = 2,2':6',2''-terpyridyl; py = pyridine; dppz = dipyrido[3,2-a:2'3'-c]phenazine) with DNA, providing detailed information about the
Fujun Li et al.
Nature communications, 6, 7843-7843 (2015-07-25)
Lithium-oxygen cells have attracted extensive interests due to their high theoretical energy densities. The main challenges are the low round-trip efficiency and cycling instability over long time. However, even in the state-of-the-art lithium-oxygen cells the charge potentials are as high
Denise Lego et al.
NMR in biomedicine, 27(7), 810-816 (2014-05-09)
Parahydrogen-induced polarization (PHIP) is a promising new tool for medical applications of MR, including MRI. The PHIP technique can be used to transfer high non-Boltzmann polarization, derived from parahydrogen, to isotopes with a low natural abundance or low gyromagnetic ratio

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service