Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44245

Sigma-Aldrich

Dodecyltrimethylammonium hydrogen sulfate

≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11N(CH3)3HSO4
CAS Number:
Molecular Weight:
325.51
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

assay

≥98.0% (T)

SMILES string

OS([O-])(=O)=O.CCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C15H34N.H2O4S/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;1-5(2,3)4/h5-15H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

KSZDKSNRUYOJHR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service