Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442240

Sigma-Aldrich

Diisodecylphenyl phosphite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H21O)2POC6H5
CAS Number:
Molecular Weight:
438.62
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

refractive index

n20/D 1.48 (lit.)

bp

176 °C/5 mmHg (lit.)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCCCCCOP(OCCCCCCCC(C)C)Oc1ccccc1

Application

Heat stabilizer for polymers

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service