Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

441600

Sigma-Aldrich

Captopril

97%

Synonym(s):

N-[(S)-3-Mercapto-2-methylpropionyl]-L-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO3S
CAS Number:
Molecular Weight:
217.29
Beilstein/REAXYS Number:
477887
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

97%

optical activity

[α]20/D −129°, c = 1.7 in ethanol

mp

104-108 °C (lit.)

SMILES string

C[C@H](CS)C(=O)N1CCC[C@H]1C(O)=O

InChI

1S/C9H15NO3S/c1-6(5-14)8(11)10-4-2-3-7(10)9(12)13/h6-7,14H,2-5H2,1H3,(H,12,13)/t6-,7+/m1/s1

InChI key

FAKRSMQSSFJEIM-RQJHMYQMSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Angiotensin converting enzyme inhibitor. Inhibits the formation of angiotensin II, a bioactive peptide that stimulates angiogenesis and increases microvessel density.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas G Tzakos et al.
Bioorganic & medicinal chemistry letters, 16(19), 5084-5087 (2006-08-08)
Enzyme-inhibitor recognition is considered one of the most fundamental aspects in the area of drug discovery. However, the molecular mechanism of this recognition process (induced fit or prebinding and adaptive selection among multiple conformers) in several cases remains unexplored. In
Ioanna Ntai et al.
Bioorganic & medicinal chemistry letters, 18(10), 3068-3071 (2007-12-22)
The naturally occurring phosphonotripeptide K-26 is a potent angiotensin converting enzyme (ACE) inhibitor containing an alpha-amino phosphonic acid analogue of tyrosine. Previous studies have demonstrated that canonical peptide analogues of K-26 are micromolar inhibitors of ACE. To ascertain the structure-activity
Guo-Xiang Li et al.
European journal of medicinal chemistry, 44(12), 4841-4847 (2009-08-21)
Antioxidant capacities of captopril (CAP), 6-mercaptopurine (6-MP) and 9-(beta-D-ribofuranosyl)-6-mercaptopurine (6-MPR) were investigated by interacting them with 2,2'-diphenyl-1-picrylhydrazyl (DPPH), galvinoxyl radical, and 2,2'-azinobis(3-ethylbenzothiazoline-6-sulfonate) cation radical (ABTS(+)(*)), and by protecting DNA and erythrocyte against 2,2'-azobis(2-amidinopropane hydrochloride) (AAPH) induced oxidation. It was found
A Ma Velázquez et al.
European journal of medicinal chemistry, 43(3), 486-500 (2007-06-22)
We present in this work the synthesis and cardiovascular effects of new methylthiomorpholine compounds and they were compared with cardiovascular drugs such as captopril, losartan and omapatrilat.
Hiroshi Enomoto et al.
Bioorganic & medicinal chemistry letters, 18(16), 4529-4532 (2008-08-05)
We studied the synthetic modification of structurally similar N-mercaptoacyl-L-proline and (4R)-N-mercaptoacylthiazolidine-4-carboxylic acid to obtain potent leukotriene A(4) (LTA(4)) hydrolase inhibitors. An N-mercaptoacyl group, (2S)-3-mercapto-2-methylpropionyl group, was effective for both scaffolds. Additional introduction of a large substituent such as 4-isopropylbenzylthio (3f)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service